Learn More
Bio X Cell Antibody, Anti-Mouse CD40, FGK4.5/FGK45, IgG2a, 50 MG
Supplier: Bio X Cell BE00162R050MG
This InVivoMAb anti-mouse CD40 antibody, clone FGK4.5/FGK45, is a rat IgG2a isotype designed for research applications.
- Targets mouse CD40
- Clone: FGK4.5/FGK45
- Isotype: Rat IgG2a
- Provided unconjugated
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.