Learn More
Bio X Cell Antibody, anti-mouse CD45RB, Clone MB23G2, 5 MG
Supplier: Bio X Cell BE0019R005MG
The HB220 antibody reacts with mouse CD45RB, a 220 kDa transmembrane protein tyrosine phosphatase expressed on peripheral B cells, naive T cells, thymocytes, and weakly on macrophages and dendritic cells. CD45RB is critical for intracellular signaling in T cells in response to antigen stimulation.
- Clone MB23G2 (HB220)
- Isotype rat IgG2a, κ
- Reactivity mouse
- Applications: in vivo anti-CD45RB tolerance induction, in vivo pre-mNK cell depletion, flow cytometry
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Immunogen: anti-immunoglobulin activated mouse B cells
- Endotoxin ≤1EU/mg (≤0.001EU/μg)
- Purity ≥95% by SDS-PAGE
- Sterility 0.2 μm filtered
- Production in animal-free facility
- Purification: Protein G
- Molecular weight 150 kDa
- Store at stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.