Learn More
Bio X Cell InVivoMAb, anti-mouse CD54, Rat IgG2b, 5 MG
Supplier: Bio X Cell BE00201R005MG
The YN1/1.7.4 monoclonal antibody reacts with mouse CD54, also known as Intercellular Adhesion Molecule-1 (ICAM-1). CD54 is a 90 kDa single-chain transmembrane glycoprotein that acts as a ligand for LFA-1 on leukocytes, facilitating transmigration into tissues. This antibody neutralizes CD54 in vivo, inhibiting CXCL1, IFNγ, and IL-17 production and neutrophil infiltration into inflammatory lesions.
- Reported applications: In vivo ICAM-1 neutralization, immunohistochemistry (frozen), ELISA
- Formulated in PBS, pH 7.0, containing no stabilizers or preservatives
- Purity: ≥95% determined by SDS-PAGE
- Endotoxin: ≤1EU/mg (≤0.001EU/µg), determined by LAL assay
- Sterility: 0.2 µm filtered
- Produced in an animal-free facility, purified by Protein G
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.