Learn More
Bio X Cell CD8, YTS 169.4, Rat IgG2b, 5 MG
Supplier: Bio X Cell BE0117R005MG
The YTS 169.4 monoclonal antibody reacts with mouse CD8α, a transmembrane glycoprotein that acts as a co-receptor for the T cell receptor (TCR) and binds to class I MHC molecules. This antibody is primarily expressed on cytotoxic T cells and exhibits depleting activity when used in vivo.
- Reactivities: Mouse
- Isotype: Rat IgG2b, κ
- Clone: YTS 169.4
- Reported applications: In vivo CD8+ T cell depletion, Western blot
- Conjugation: Unconjugated
- Immunogen: CBA mouse thymocytes
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.