Learn More
Bio X Cell InVivoMAb anti-mouse CD8 YTS 169.4, 100 MG
Supplier: Bio X Cell BE0117R100MG
The YTS 169.4 monoclonal antibody reacts with mouse CD8α. CD8 is a transmembrane glycoprotein that functions as a co-receptor for the T cell receptor (TCR) and is primarily found on cytotoxic T cells.
- Reacts with mouse CD8α
- Isotype: Rat IgG2b, κ
- Unconjugated
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C. Do not freeze
- Applications: in vivo CD8+ T cell depletion, Western blot
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.