Learn More
Bio X Cell Anti-Mouse CD8 Alpha, Clone 2.43, 5 MG
Supplier: Bio X Cell BE0061R005MG
The 2.43 monoclonal antibody reacts with mouse CD8α. The CD8 antigen is a transmembrane glycoprotein that acts as a co-receptor for the T cell receptor (TCR), primarily expressed on cytotoxic T cells. This antibody exhibits depleting activity when used in vivo.
- Target: mouse CD8α
- Clone: 2.43
- Isotype: Rat IgG2b, κ
- Applications: in vivo CD8+ T cell depletion, Western blot
- Purification method: Protein G
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin ≤1EU/mg, purity ≥95%, 0.2 μm sterile filtration
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.