Learn More
Bio X Cell CD8 alpha, Clone 2.43, Unconjugated, 50 MG
Supplier: Bio X Cell BE0061R050MG
This monoclonal antibody, clone 2.43, targets mouse CD8α, a T cell co-receptor found on cytotoxic T cells, thymocytes, NK cells, and dendritic cell subsets. It demonstrates depleting activity in vivo.
- Isotype: Rat IgG2b, κ
- Reactivity: Mouse
- Immunogen: Mouse CTL clone L3
- Applications: In vivo CD8+ T cell depletion, Western blot
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95% (SDS-PAGE)
- Sterility: 0.2 μm filtration
- Production: Purified from animal-free cell culture
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
- Conjugation: Unconjugated
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.