Learn More
Bio X Cell InVivoMAb anti-Mouse CD8alpha, 53-6.7 IgG2a, 25 MG
Supplier: Bio X Cell BE00041R025MG
The 53-6.7 monoclonal antibody reacts with mouse CD8α. CD8 is a transmembrane glycoprotein that functions as a co-receptor for the T cell receptor (TCR) and binds to class I MHC molecules presented by antigen-presenting cells (APC). It is primarily expressed on cytotoxic T cells, thymocytes, natural killer cells, and some dendritic cell subsets. This antibody has in vivo depleting activity.
- Isotype: Rat IgG2a, κ
- Purity: ≥95%, determined by SDS-PAGE
- Endotoxin: ≤1EU/mg (≤0.001EU/μg), determined by LAL assay
- Applications: In vivo CD8+ T cell depletion, Immunofluorescence, Flow cytometry, Western blot
- Formulation: PBS, pH 6.5; contains no stabilizers or preservatives
- Purification: Protein G
- Storage: Store at stock concentration at 4°C. Do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.