Learn More
Bio X Cell Anti-Mouse CD8alpha, Clone 53-6.7, IgG2a, 100 MG
Supplier: Bio X Cell BE00041R100MG
The 53-6.7 monoclonal antibody reacts with mouse CD8α. CD8 is a transmembrane glycoprotein that acts as a co-receptor for the T cell receptor (TCR) and binds to class I MHC molecules. This antibody is primarily expressed on cytotoxic T cells and exhibits depleting activity in vivo.
- Reactivities: mouse
- Host species: rat
- Isotype: IgG2a, κ
- Unconjugated
- Formulation: PBS, pH 6.5, contains no stabilizers or preservatives
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Purity: ≥95% (determined by SDS-PAGE)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Applications: in vivo CD8+ T cell depletion, immunofluorescence, flow cytometry, western blot
- Storage: store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.