Learn More
Bio X Cell Anti-Mouse CTLA-4, Clone 9D9, IgG2b, 5 MG
Supplier: Bio X Cell BE0164R005MG
The 9D9 monoclonal antibody reacts with mouse CTLA-4 (cytotoxic T lymphocyte antigen-4), also known as CD152. This antibody negatively regulates cell-mediated immune responses, playing roles in immunological tolerance, thymocyte development, and protective immunity.
- Isotype: mouse IgG2b
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Produced in an animal-free facility
- Purification method: Protein A
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.