Learn More
Bio X Cell Antibody, Anti-Mouse CTLA-4, Clone 9D9, 25 MG
Supplier: Bio X Cell BE0164R025MG
The 9D9 monoclonal antibody targets mouse CTLA-4 (cytotoxic T lymphocyte antigen-4), also known as CD152. CTLA-4 is a 33 kDa cell surface receptor expressed on activated T and B lymphocytes. It negatively regulates cell-mediated immune responses and plays roles in immunological tolerance, thymocyte development, and regulation of protective immunity.
- Reacts with mouse CTLA-4 (CD152)
- Clone: 9D9
- Isotype: Mouse IgG2b
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Purity: ≥95% by SDS-PAGE
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Sterility: 0.2 μm filtration
- Purification: Protein A, from animal-free cell culture supernatant
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.