Learn More
Bio X Cell Antibody, anti-mouse CTLA-4, Clone 9H10, 100 MG
Supplier:  Bio X Cell BP0131R100MG
Encompass_Preferred
The 9H10 monoclonal antibody reacts with mouse CTLA-4 (CD152), a cell surface receptor that negatively regulates cell-mediated immune responses. This antibody promotes T cell co-stimulation by blocking CTLA-4 binding to B7 co-receptors, allowing for CD28 binding, which is critical in immune down-regulation and explored as a cancer treatment target.
  • Monoclonal antibody (clone 9H10)
  • Reacts with mouse CTLA-4 (CD152)
  • Promotes T cell co-stimulation by blocking CTLA-4 binding
  • InVivoMAb format for in vivo functional studies
  • Purified from cell culture supernatant in an animal-free facility
  • Sterility ensured by 0.2 μm filtration
  • Low endotoxin level (≤1EU/mg)
  • High purity (≥95%)
  • Contains no stabilizers or preservatives
Catalog No. 50-139-3603
Add to Cart
Request a Quote