Learn More
Bio X Cell Antibody, Mouse MHC (H-2), M1/42.3.9.8, 5 MG
Supplier: Bio X Cell BE0077R005MG
The M1/42.3.9.8 monoclonal antibody reacts with the mouse H-2 MHC class I alloantigen (all haplotypes). MHC class I antigens are heterodimers consisting of one alpha chain (44 kDa) associated with β2 microglobulin (11.5 kDa). The antigen is expressed by all nucleated cells at varying levels. MHC Class I molecules present endogenously synthesized antigenic peptides to CD8 T cells.
- Validated for ex vivo blocking, MHC-I immunopeptidomics, immunoprecipitation, flow cytometry, and immunofluorescence
- Formulated in PBS, pH 7.0, contains no stabilizers or preservatives
- Purity ≥95% (SDS-PAGE)
- Endotoxin ≤1EU/mg (≤0.001EU/μg) by LAL assay
- Sterility: 0.2 μm filtered
- Produced in animal-free facility, purified by Protein G
- Molecular weight: 150 kDa
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.