Learn More
Bio X Cell Antibody, anti-mouse/human CD44, Clone IM7, Rat IgG2b, 100 MG
Supplier: Bio X Cell BE0039R100MG
This monoclonal antibody, clone IM7, reacts with human and mouse CD44, an 80-95 kDa glycoprotein found on various cell types. CD44 acts as an adhesion molecule crucial for lymphocyte activation, homing, and hematopoiesis, and is also a receptor for hyaluronic acid. Implicated in tumor metastasis, this antibody effectively neutralizes CD44 in vivo.
- Reactivities: Mouse, human
- Isotype: Rat IgG2b,
- Unconjugated
- Immunogen: Dexamethasone-induced myeloid leukemia M1 cells
- Formulation: PBS, pH 7.0 (no stabilizers or preservatives)
- Endotoxin: 1EU/mg
- Purity: 95%
- Sterility: 0.2 m filtered
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: 4C, do not freeze
- Applications: In vivo and in vitro CD44 neutralization
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.