Learn More
Bio X Cell Monoclonal Antibody, TGF-beta, Mouse IgG1, 25 MG
Supplier: Bio X Cell BE0057R025MG
The InVivoMAb anti-mouse/human/rat/monkey/hamster/canine/bovine TGF-β is a monoclonal antibody designed for both in vivo and in vitro TGFβ neutralization, and Western blot applications.
- Host: Mouse
- Isotype: IgG1, κ
- Immunogen: Bovine TGFβ isoform 2
- Applications: in vivo TGFβ neutralization, in vitro TGFβ neutralization, Western blot
- Storage: Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.