Learn More
Bio X Cell InVivoMAb Antibody, anti-TGF-beta, Mouse Host, 5 MG
Supplier: Bio X Cell BE0057R005MG
This InVivoMAb antibody is designed for the neutralization and detection of TGFβ across multiple species including mouse, human, rat, monkey, hamster, canine, and bovine. It is supplied as a 5 mg package.
- Applications include in vivo and in vitro TGFβ neutralization, and Western blot.
- Antibody host is mouse.
- Reacts with bovine, canine, hamster, human, monkey, mouse, and rat species.
- Isotype is IgG1, κ.
- Immunogen is bovine TGFβ isoform 2.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.