Learn More
Bio X Cell Antibody, Mouse IFN gamma, XMG1.2, 100 MG
Supplier: Bio X Cell BE0055R100MG
The XMG1.2 monoclonal antibody reacts with mouse IFNγ (interferon gamma), a 20 kDa soluble pleiotropic cytokine. This antibody is a neutralizing antibody and plays an important role in the activation, growth, and differentiation of various immune and non-hematopoietic cell types.
- Isotype: Rat IgG1, κ
- Immunogen: Recombinant mouse IFNγ
- Applications: In vivo IFNγ neutralization, in vitro IFNγ neutralization, ELISPOT, Flow cytometry, Western blot
- Formulation: PBS + 0.01% Tween, pH 8.0, no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.