Learn More
Bio X Cell Antibody, Mouse IFN-gamma R4-6A2, IgG1, 5 MG
Supplier: Bio X Cell BE0054R005MG
The R4-6A2 monoclonal antibody reacts with mouse IFNγ (interferon gamma), a 20 kDa soluble pleiotropic cytokine. This antibody neutralizes both natural and recombinant IFNγ, which plays a crucial role in immunoregulatory, anti-proliferative, anti-viral, and proinflammatory activities.
- Produced in an animal-free facility.
- Endotoxin level ≤1 EU/mg.
- Purity ≥95%.
- Formulated in PBS, pH 8.0, with no stabilizers or preservatives.
- Sterility by 0.2 μm filtration.
- Purified by Protein G.
- Molecular weight of 150 kDa.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.