Learn More
Bio X Cell Antibody, Anti-Mouse IFN XMG1.2, Rat IgG1, Isotype, 5 MG
Supplier: Bio X Cell BE0055R005MG
The XMG1.2 monoclonal antibody targets mouse IFN (interferon gamma), a 20 kDa pleiotropic cytokine. Primarily produced by activated lymphocytes, IFN is vital for immunoregulatory, anti-proliferative, anti-viral, and proinflammatory responses. This neutralizing antibody modulates IFN's role in immune cell activation, growth, and differentiation, as well as its influence on cytokine production and MHC class I/II antigen expression.
- Reacts with mouse IFN, Clone XMG1.2
- Rat IgG1, isotype
- Immunogen: Recombinant mouse IFN
- Formulation: PBS + 0.01% Tween, pH 8.0, no stabilizers or preservatives
- Endotoxin: 1EU/mg
- Purity: 95% (SDS-PAGE)
- Sterility: 0.2 m filtered
- Produced in an animal-free facility via Protein G purification
- Molecular weight: 150 kDa
- Applications include: in vivo/in vitro IFN neutralization, ELISPOT, Flow cytometry, Western blot, and in vitro Organoids/Organ-on-Chip
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.