Learn More
Bio X Cell Antibody, anti-mouse IFN gamma, XMG1.2, 25 MG
Supplier: Bio X Cell BE0055R025MG
The XMG1.2 monoclonal antibody reacts with mouse IFNγ (interferon gamma), a 20 kDa soluble pleiotropic cytokine. It exerts immunoregulatory, anti-proliferative, anti-viral, and proinflammatory activities, playing an important role in activation, growth, and differentiation of various cell types. The XMG1.2 antibody is a neutralizing antibody.
- In vivo and in vitro IFNγ neutralization
- ELISPOT, Flow cytometry, and Western blot applications
- Formulated in PBS + 0.01% Tween, pH 8.0, no stabilizers or preservatives
- Purified using Protein G
- Purity ≥95% (SDS-PAGE)
- Endotoxin ≤1EU/mg (LAL assay)
- Sterile filtered (0.2 μm)
- Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.