Learn More
Bio X Cell Antibody, Mouse IFNAR-1, MAR1-5A3, IgG1, 25 MG
Supplier: Bio X Cell BE0241R025MG
The MAR1-5A3 monoclonal antibody targets mouse IFNAR-1 (IFN alpha/beta receptor subunit 1). It inhibits Type I IFN receptor signaling both in vitro and in vivo.
- Clonality: Monoclonal
- Isotype: Mouse IgG1, κ
- Immunogen: Extracellular domain of mouse IFNAR-1
- Purity: ≥95% (determined by SDS-PAGE)
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Purification: Protein G
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Applications: In vivo and in vitro IFNAR-1 blockade, Western blot, Flow Cytometry, ELISA
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.