Learn More
Bio X Cell anti-mouse IFNAR-1, Clone MAR1-5A3, Unconjugated, 5 MG
Supplier: Bio X Cell BE0241R005MG
The MAR1-5A3 monoclonal antibody reacts with mouse IFNAR-1 (IFN α/β receptor subunit 1), a subunit of the Type I IFN Receptor complex. This antibody has been shown to inhibit Type I IFN receptor signaling both in vitro and in vivo, affecting anti-viral, anti-microbial, anti-tumor, and autoimmune responses.
- Clone: MAR1-5A3
- Reactivities: Mouse
- Isotype: Mouse IgG1, κ
- Conjugation: Unconjugated
- Immunogen: Extracellular domain of mouse IFNAR-1
- Reported applications: In vivo/in vitro IFNAR-1 blockade, Western blot, Flow Cytometry, ELISA
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.