Learn More
Bio X Cell InVivoMAb Anti-mouse IL-1 alpha ALF-161, 5 MG
Supplier: Bio X Cell BE0243R005MG
The ALF-161 monoclonal antibody reacts with precursor, secreted, and membrane-associated forms of mouse IL-1α (interleukin 1 alpha). It has been shown to neutralize the bioactivity of natural or recombinant IL-1α, which is a 17 kDa pro-inflammatory cytokine involved in a wide range of immune and inflammatory responses.
- Reacts with precursor, secreted, and membrane-associated forms of mouse IL-1α
- Neutralizes bioactivity of natural or recombinant IL-1α
- Purified by Protein A from cell culture supernatant in an animal-free facility
- Formulated in PBS, pH 7.0, with no stabilizers or preservatives
- Purity ≥95% (SDS-PAGE) and Endotoxin ≤1EU/mg
- Suitable for in vivo and in vitro IL-1α neutralization
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.