Learn More
Bio X Cell InVivoMAb Antibody, anti-IL-1B, Clone B122, 5 MG
Supplier: Bio X Cell BE0246R005MG
The B122 monoclonal antibody targets both precursor and mature secreted forms of mouse and rat IL-1β, a 17 kDa pro-inflammatory cytokine primarily produced by monocytes. This antibody neutralizes the bioactivity of natural or recombinant IL-1β and is involved in cellular activities such as proliferation, differentiation, apoptosis, and T and B lymphocyte activation.
- Formulated in PBS, pH 7.0; no stabilizers or preservatives
- Endotoxin ≤1 EU/mg (≤0.001 EU/μg) by LAL assay
- Purity ≥95% by SDS-PAGE
- Sterility via 0.2 μm filtration
- Produced from animal-free cell culture supernatant
- Protein G purified
- Molecular weight is 150 kDa
- Store at 4°C; do not freeze
- Validated for in vivo/in vitro IL-1β neutralization, ELISA
- Immunogen: recombinant mouse IL-1β
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.