Learn More
Bio X Cell Anti-mouse IL-10, Clone JES5-2A5, 5 MG
Supplier: Bio X Cell BE0049R005MG
The JES5-2A5 monoclonal antibody targets mouse IL-10, a 17-21 kDa cytokine involved in immunoregulation and inflammation. It can neutralize the bioactivity of natural or recombinant IL-10.
- Isotype: Rat IgG1, κ
- Reactivity: Mouse
- Immunogen: Recombinant mouse IL-10
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg), determined by LAL assay
- Purity: ≥95%, determined by SDS-PAGE
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C, do not freeze
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Reported applications: In vivo IL-10 neutralization, in vitro IL-10 neutralization
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.