Learn More
Bio X Cell 1B1.3A, Mouse IL-10R, Rat IgG1, 5 MG
Supplier: Bio X Cell BE0050R005MG
The 1B1.3A monoclonal antibody reacts with mouse IL-10R (CD210), a class II cytokine receptor expressed by various cell types. This neutralizing antibody blocks human IL-10 binding, cross-reacting with mouse IL-10R, and mediates immunoregulation and inflammation.
- In vivo and in vitro blocking of IL-10/IL-10R signaling
- Flow cytometry and Western blot applications
- Rat IgG1, κ isotype
- Immunogen: Recombinant ligand-binding domain of mouse IL-10R
- Purified by Protein G
- Formulated in PBS, pH 6.5, 0.01% Tween, without stabilizers or preservatives
- Purity ≥95% by SDS-PAGE
- Endotoxin ≤1EU/mg
- Sterility: 0.2 μm filtration
- Molecular weight: 150 kDa
- Store at stock concentration at 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.