Learn More
Bio X Cell IL-12 p40 C17.8, Rat IgG2a, 0.5 MG
Supplier: Bio X Cell BE0051R005MG
The C17.8 antibody reacts with mouse p40, also known as IL-12β. This 40 kDa subunit is part of IL-12 and IL-23, heterodimeric cytokines involved in T lymphocyte differentiation, IFNγ production, and NK cell cytotoxicity. The C17.8 antibody neutralizes both IL-12 and IL-23 bioactivity.
- Host species: Rat
- Target species: Mouse
- Isotype: Rat IgG2a, κ
- Purification method: Protein G
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/μg)
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 μm filtration
- Applications include: in vivo IL-12p40 neutralization, p40 affinity chromatography, Immunoprecipitation, ELISA, Flow cytometry, Western blot
- Storage: Store at stock concentration at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.