Learn More
Bio X Cell Antibody, anti-mouse IL-12 p40, 25 MG
Supplier: Bio X Cell BE0051R025MG
The C17.8 antibody targets mouse IL-12 p40, a subunit of both IL-12 and IL-23, and neutralizes the bioactivity of both cytokines, which are involved in T lymphocyte differentiation, IFNγ production, and NK cell cytotoxicity.
- Host species: Rat
- Isotype: Rat IgG2a, κ
- Immunogen: Recombinant mouse IL-12 p70
- Purity: ≥95%
- Endotoxin: ≤1EU/mg
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
- Purification method: Protein G
- Production: Purified from cell culture supernatant in an animal-free facility
- Applications: in vivo IL-12p40 neutralization, p40 affinity chromatography, immunoprecipitation, ELISA, flow cytometry, western blot
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.