Learn More
Bio X Cell Antibody, Anti-Mouse IL-12 p40, 5 MG
Supplier: Bio X Cell BP0051R005MG
The C17.8 antibody reacts with mouse p40, a 40 kDa subunit of both IL-12 and IL-23. This antibody neutralizes both IL-12 and IL-23 bioactivity.
- Isotype: Rat IgG2a, κ
- Reactivities: Mouse
- Clone: C17.8
- Conjugation: Unconjugated
- Immunogen: Recombinant mouse IL-12 p70
- Reported applications: In vivo IL-12p40 neutralization, p40 affinity chromatography, immunoprecipitation, ELISA, flow cytometry, western blot
- Formulation: PBS, pH 7.0
- Purity: ≥95%
- Endotoxin: ≤0.5EU/mg
- Aggregation: <5%
- Storage: Store at 4°C; do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.