Learn More
Bio X Cell Antibody, Anti-Mouse/Rat IL-17A, Clone 17F3, 25 MG
Supplier: Bio X Cell BE0173R025MG
The 17F3 monoclonal antibody targets mouse IL-17A, a 15-20 kDa cytokine crucial in immunoregulation and inflammation. This antibody neutralizes IL-17A in vivo, playing an important role in anti-microbial and chronic inflammation research, as well as studies on autoimmune diseases.
- Reactivity: mouse, rat
- Isotype: mouse IgG1, κ
- Immunogen: mouse IL-17A cross-linked to OVA
- Reported applications: in vivo IL-17A neutralization, ELISA
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤1EU/mg (≤0.001EU/µg)
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 µm filtration
- Purification method: Protein G
- Molecular weight: 150 kDa
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.