Learn More
Bio X Cell Antibody, Mouse/Rat IL-17A, Clone 17F3, 25 MG
Supplier: Bio X Cell BP0173R025MG
The 17F3 monoclonal antibody reacts with mouse IL-17A, a cytokine involved in immunoregulation and inflammation. This antibody neutralizes IL-17A in vivo, playing an important role in anti-microbial and chronic inflammation research, and also in the study of autoimmune diseases like rheumatoid arthritis and psoriasis.
- Clone: 17F3
- Isotype: Mouse IgG1, κ
- Reactivity: Mouse, Rat
- Applications: In vivo IL-17A neutralization, ELISA
- Immunogen: Mouse IL-17A cross-linked to OVA
- Formulation: PBS, pH 7.0; contains no stabilizers or preservatives
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Storage: Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.