Learn More
Bio X Cell Antibody, Mouse/Rat IL-17A, Clone 17F3, 5 MG
Supplier:  Bio X Cell BP0173R005MG
Encompass_Preferred
The 17F3 monoclonal antibody targets mouse IL-17A, a 15-20 kDa cytokine crucial for immunoregulation and inflammation. It effectively neutralizes IL-17A in vivo, making it valuable for research in autoimmune diseases, anti-microbial responses, and chronic inflammation. This unconjugated product is purified from cell culture supernatant in an animal-free facility, offering high purity (≥95%) and ultra-low endotoxin levels (≤0.5EU/mg).
  • Reacts with mouse/rat IL-17A cytokine
  • Suitable for in vivo neutralization and ELISA applications
  • Purified from cell culture supernatant in an animal-free facility
  • High purity (≥95%) and ultra-low endotoxin levels (≤0.5EU/mg)
  • Formulated in PBS, pH 7.0; free of stabilizers and preservatives
Catalog No. 50-139-3612