Learn More
Bio X Cell Antibody, anti-mouse IL-2, S4B6-1 IgG2a, 5 MG
Supplier: Bio X Cell BE00431R005MG
InVivoMAb anti-mouse IL-2 is an antibody solution designed for in vivo applications, specifically for IL-2 neutralization and receptor stimulation.
- Used for in vivo IL-2 neutralization
- Used for in vivo IL-2 receptor stimulation (as a complex with IL-2)
- Store at 4°C; do not freeze
- Shipped at 4°C with blue ice
- Host species: Rat
- Target species: Mouse
- Isotype: IgG2a
- Immunogen: Recombinant mouse IL-2
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.