Learn More
Bio X Cell anti-mouse IL-4, Rat IgG1, Unconjugated, 5 MG
Supplier: Bio X Cell BE0045R005MG
This antibody is specifically developed to target mouse IL-4, offering reliable performance for various research applications. It is provided in an unconjugated format, purified using Protein G for high specificity.
- Antibody host: Rat
- Species reactivity: Mouse
- Isotype: IgG1, κ
- Immunogen: Partially purified native mouse IL-4
- Applications: In vivo IL-4 neutralization, in vitro IL-4 neutralization, in vivo IL-4 receptor stimulation (as a complex with IL-4), Flow cytometry, Western blot
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.