Learn More
Bio X Cell Anti-mouse IL-4 11B11, Rat IgG1, 5 MG
Supplier:  Bio X Cell BP0045R005MG
Encompass_Preferred
The 11B11 monoclonal antibody reacts with mouse IL-4 (interleukin-4), a multifunctional 14 kDa cytokine. It signals through the IL-4Rα, stimulating activated B and T lymphocyte proliferation and differentiation of B cells into plasma cells. It also induces B cell class switching to IgE and up-regulates MHC class II production.
  • Reacts with mouse IL-4
  • Monoclonal antibody, clone 11B11
  • Rat IgG1, κ isotype
  • Purity: ≥95%
  • Endotoxin level: ≤0.5EU/mg
  • Validated for in vivo and in vitro neutralization, flow cytometry, and Western blot
Catalog No. 50-139-3533