Learn More
Bio X Cell Monoclonal Antibody, anti-Mouse IL-6, MP5-20F3, 25 MG
Supplier: Bio X Cell BE0046R025MG
The MP5-20F3 monoclonal antibody reacts with mouse IL-6, a 21-28 kDa cytokine expressed by various cell types. This antibody neutralizes the bioactivity of natural or recombinant IL-6, influencing antigen-specific immune responses, inflammatory responses, and neuronal development.
- Target: mouse IL-6
- Clone: MP5-20F3
- Isotype: rat IgG1, κ
- Reactivities: mouse
- Reported applications: in vivo IL-6 neutralization, in vitro IL-6 neutralization
- Immunogen: recombinant mouse IL-6
- Endotoxin: ≤1EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Production: purified from cell culture supernatant in an animal-free facility
- Purification method: Protein G
- Molecular weight: 150 kDa
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.