Learn More
Bio X Cell anti-mouse IL-6, MP5-20F3, Rat IgG1, 5 MG
Supplier: Bio X Cell BE0046R005MG
The MP5-20F3 monoclonal antibody reacts with mouse IL-6, a cytokine influencing immune responses, inflammation, and neuronal development. It neutralizes natural or recombinant IL-6 bioactivity.
- Clone: MP5-20F3
- Reactivities: Mouse
- Isotype: Rat IgG1, κ
- Immunogen: Recombinant mouse IL-6
- Formulation: PBS, pH 7.0 (contains no stabilizers or preservatives)
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg) via LAL assay
- Purity: ≥95% via SDS-PAGE
- Sterility: 0.2 μm filtration
- Production: Purified from cell culture supernatant in an animal-free facility
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store the antibody solution at the stock concentration at 4°C. Do not freeze
- Applications: In vivo and in vitro IL-6 neutralization
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.