Learn More
Bio X Cell InVivoMAb Antibody, anti-mouse LAG-3, C9B7W Clone, Rat IgG1, Sterile, 5 MG
Supplier: Bio X Cell BE0174R005MG
This InVivoMAb anti-mouse LAG-3 antibody, with clone C9B7W, is a rat IgG1, kappa isotype. It is specifically designed for in vivo and in vitro LAG-3 neutralization, flow cytometry, and Western blot applications. Purified from cell culture supernatant, this antibody is provided sterile and exhibits high purity, making it suitable for sensitive research.
- Reacts with mouse LAG-3 (CD223)
- Isotype: rat IgG1, κ
- Recommended for in vivo and in vitro LAG-3 neutralization, flow cytometry, Western blot
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin level: ≤1EU/mg
- Purity: ≥95% by SDS-PAGE
- Sterility: 0.2 µm filtration
- Production from animal-free facility
- Purification method: protein G
- Molecular weight: 150 kDa
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.