Learn More
Bio X Cell Antibody, anti-LCMV Nucleoprotein, Clone VL-4, Rat IgG2a, Immunofluorescence, 5 MG
Supplier: Bio X Cell BE0106R005MG
This antibody reacts with lymphocytic choriomeningitis virus (LCMV) nucleoprotein (NP), a 63 kDa structural protein. It was generated by fusing spleen cells from an LCMV strain WE immunized F1 rat with the YM3 myeloma cell line. The antibody stains LCMV-infected cells internally with no surface staining and does not react with vaccinia, vesicular stomatitis, or influenza virus-infected cells.
- Clone VL-4.
- Isotype rat IgG2a, .
- Immunogen LCMV strain WE.
- Reported applications include immunofluorescence and flow cytometry.
- Formulated in PBS, pH 7.0, without stabilizers or preservatives.
- Endotoxin level is 1 EU/mg.
- Purity is 95%.
- Sterility is 0.2 m filtered.
- Purified in an animal-free facility.
- Purification by protein G.
- Molecular weight is 150 kDa.
- Store at 4C; do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.