Learn More
Bio X Cell Antibody, Anti-Mouse Ly6G, Rat IgG2a, 25 MG
Supplier: Bio X Cell BE00751R025MG
The 1A8 monoclonal antibody reacts specifically with mouse Ly6G, a 21-25 kDa member of the Ly-6 superfamily of GPI-anchored cell surface proteins involved in cell signaling and adhesion. Unlike the RB6-8C5 antibody, 1A8 does not have reported cross-reactivity with Ly6C.
- Reacts specifically with mouse Ly6G, no cross-reactivity with Ly6C
- Isotype: Rat IgG2a, κ
- Purity: ≥95%
- Endotoxin: ≤1EU/mg
- Sterility: 0.2 μm filtration
- Purification: Protein G
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Molecular Weight: 150 kDa
- Applications: in vivo neutrophil/MDSC depletion, immunofluorescence, immunohistochemistry, flow cytometry
- Clone: 1A8
- Unconjugated
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.