Learn More
Bio X Cell Antibody, Ly6G/Ly6C (Gr-1), Clone RB6-8C5, 5 MG
Supplier: Bio X Cell BE0075R005MG
InVivoMAb anti-mouse Ly6G/Ly6C (Gr-1) is a rat monoclonal antibody (clone RB6-8C5) targeting mouse Ly6G and Ly6C. This antibody is suitable for various applications including in vivo depletion of Gr-1+ myeloid cells, flow cytometry, and immunohistochemistry.
- Clone: RB6-8C5
- Host species: Rat
- Target: Mouse Ly6G and Ly6C (Gr-1)
- Applications: in vivo depletion of Gr-1+ myeloid cells, flow cytometry, immunohistochemistry (paraffin), immunohistochemistry (frozen)
- Purification method: Protein G
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Storage conditions: Store at stock concentration at 4°C, do not freeze
- Isotype: Rat IgG2b, κ
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.