Learn More
Bio X Cell Antibody, Ly6G/Ly6C (Gr-1), Rat IgG2b, 25 MG
Supplier: Bio X Cell BE0075R025MG
This antibody is an InVivoMAb anti-mouse Ly6G/Ly6C (Gr-1) monoclonal antibody. It is designed for applications requiring detection or depletion of Ly6G/Ly6C (Gr-1) antigens in mouse models.
- Applications include in vivo depletion of Gr-1+ myeloid cells, flow cytometry, immunohistochemistry (paraffin), and immunohistochemistry (frozen).
- Antibody host: Rat
- Species reactivity: Mouse
- Isotype: IgG2b, κ
- Immunogen: Mouse granulocytes
- Storage temperature: 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.