Learn More
Bio X Cell Bioxcell Monoclonal Antibody, anti-mouse NK1.1, PK136, IgG2a, 5 MG
Supplier: Bio X Cell BE0036R005MG
The PK136 monoclonal antibody targets mouse NK1.1 (CD161b/CD161c, KLRB1, NKR-P1A, Ly-55), a type II integral membrane glycoprotein crucial for NK cell activation and differentiation. It also plays roles in IFN-γ production and cytotoxic granule release.
- Clone: PK136
- Reactivity: mouse
- Isotype: mouse IgG2a, κ
- Unconjugated
- Purity: ≥95% (determined by SDS-PAGE)
- Endotoxin: ≤1 EU/mg (determined by LAL assay)
- Applications: in vivo NK cell depletion, flow cytometry
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.