Learn More
Bio X Cell Antibody, Anti-Mouse NK1.1, PK136, Unconjugated, 25 MG
Supplier: Bio X Cell BE0036R025MG
The PK136 monoclonal antibody reacts with mouse NK1.1, also known as CD161b/CD161c, KLRB1, NKR-P1A, and Ly-55. NK1.1 is a type II integral membrane glycoprotein with a C-type lectin domain, encoded by the Klrb1c/NKR-P1C gene. It plays roles in NK cell activation and differentiation, IFN-γ production, cytotoxic granule release, and is involved in Th2 cell generation.
- Isotype: Mouse IgG2a, κ
- Conjugation: Unconjugated
- Immunogen: Mouse spleen & bone marrow NK1+ cells
- Applications: In vivo NK cell depletion, flow cytometry
- Formulation: PBS, pH 7.0; no stabilizers/preservatives
- Endotoxin: ≤1 EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Production: Animal-free facility; purified from cell culture supernatant
- Purification: Protein A
- Molecular weight: 150 kDa
- Storage: Store at 4°C; do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.