Learn More
Bio X Cell Antibody, Clone PK136, Mouse IgG2a, 100 MG
Supplier: Bio X Cell BE0036R100MG
The PK136 monoclonal antibody reacts with mouse NK1.1, a type II integral membrane glycoprotein that plays roles in NK cell activation and differentiation, IFN-γ production, and cytotoxic granule release. It is predominantly expressed as a disulfide-linked homodimer on NK cells and NK-T cells.
- Clone: PK136
- Host: Mouse
- Target: Mouse NK1.1 (CD161b/CD161c, KLRB1, NKR-P1A, Ly-55)
- Isotype: Mouse IgG2a, κ
- Molecular weight: 150 kDa
- Purity: ≥95% (determined by SDS-PAGE)
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg), determined by LAL assay
- Formulation: PBS, pH 7.0, contains no stabilizers or preservatives
- Storage: Store at stock concentration at 4°C. Do not freeze.
- Purification: Purified from cell culture supernatant in an animal-free facility by Protein A.
- Applications: In vivo NK cell depletion, flow cytometry
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.