Learn More
Bio X Cell Antibody, anti-mouse NK1.1, PK136, IgG2a, 5 MG
Supplier: Bio X Cell BP0036R005MG
The PK136 monoclonal antibody reacts with mouse NK1.1, a type II integral membrane glycoprotein involved in NK cell activation, differentiation, IFN-γ production, and cytotoxic granule release. It is expressed as a disulfide-linked homodimer on NK cells and NK-T cells.
- Clone: PK136
- Host: Mouse
- Target: Mouse NK1.1 (CD161b/CD161c, KLRB1, NKR-P1A, Ly-55)
- Isotype: Mouse IgG2a, κ
- Conjugation: Unconjugated
- Applications: In vivo NK cell depletion, flow cytometry
- Purification: Protein A
- Formulation: PBS, pH 7.0, no stabilizers or preservatives
- Endotoxin: ≤0.5 EU/mg
- Purity: ≥95%
- Sterility: 0.2 μm filtration
- Aggregation: <5%
- Production: Animal-free facility
- Storage: 4°C, do not freeze
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.