Learn More
Bio X Cell Mouse OX40 (CD134), Clone OX-86, 25 MG
Supplier: Bio X Cell BP0031R025MG
The OX-86 monoclonal antibody reacts with mouse OX-40 (CD134), a 50 kDa type I membrane glycoprotein that is part of the TNF receptor superfamily. It is expressed on various activated T cells and neutrophils, providing a costimulatory signal to antigen-reacting T cells. This agonistic antibody has demonstrated efficacy in delaying tumor growth in vivo.
- Reacts with mouse OX-40 (CD134)
- Clone: OX-86
- Isotype: Rat IgG1, κ
- Applications: In vivo and in vitro OX40 activation, Western blot
- Purified by Protein G
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Storage: 4°C; do not freeze
- Endotoxin level: ≤0.5EU/mg
- Aggregation: <5%
- Purity: ≥95%
- Sterility: 0.2 µm filtration
- Produced in an animal-free facility from cell culture supernatant
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.