Learn More
Bio X Cell Antibody, Anti-Mouse OX40L, Rat IgG2b, 50 MG
Supplier: Bio X Cell BE00331R050MG
This antibody is an InVivoMAb anti-mouse OX40L (CD134L) designed for in vivo blocking of OX40/OX40L signaling and in vitro OX40L neutralization.
- Antibody host: rat
- Species reactivity: mouse
- Antibody isotype: IgG2b, κ
- Immunogen: rat NRK-52E cells transfected with mouse OX40L
- Storage temperature: 4°C; do not freeze
- Shipping temperature: 4°C with blue ice
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.