Learn More
Bio X Cell InVivoPlus anti-mouse PD-1 Clone 29F.1A12, 25 MG
Supplier: Bio X Cell BP0273R025MG
The Bio X Cell InVivoPlus anti-mouse PD-1 (CD279), Clone 29F.1A12 monoclonal antibody reacts with mouse PD-1 (CD279). This ultra high-quality version blocks PD-1 ligand binding in vivo, supporting research in anti-PD-1 immunotherapies.
- Isotype: Rat IgG2a
- Reactivities: Mouse
- Immunogen: Recombinant PD-1-Ig fusion protein
- Reported applications: In vivo PD-1/PD-L signaling blocking, in vitro PD-1 neutralization, IHC (frozen), IF, Western blot, Flow cytometry, in vitro Organoids/Organ-on-Chip
- Formulation: PBS, pH 7.0; no stabilizers or preservatives
- Endotoxin: ≤1 EU/mg (≤0.001 EU/μg)
- Purity: ≥95% (by SDS-PAGE)
- Sterility: 0.2 μm filtration
- Production: Purified from animal-free cell culture supernatant
- Purification: Protein G
- Molecular weight: 150 kDa
- Storage: Store at stock concentration at 4°C; do not freeze.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.