Learn More
Bio X Cell InVivoMAb, anti-mouse PD-1, 29F.1A12, 25 MG
Supplier: Bio X Cell BE0273R025MG
The 29F.1A12™ monoclonal antibody reacts with mouse PD-1 (programmed death-1), also known as CD279. PD-1 is a 50-55 kDa cell surface receptor encoded by the Pdcd1 gene, belonging to the CD28 family of the Ig superfamily. This antibody blocks the binding of PD-1 to its ligands in vivo, playing key roles in peripheral tolerance and prevention of autoimmune disease.
- Reacts with mouse PD-1 (CD279)
- Blocks binding of PD-1 to its ligands in vivo
- Purified by Protein G
- Formulated in PBS, pH 7.0, without stabilizers or preservatives
- Endotoxin level ≤1EU/mg
- Purity ≥95%
- Sterility by 0.2 μm filtration
- Molecular Weight: 150 kDa
- Purified from cell culture supernatant in an animal-free facility
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.